Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apoo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114243.1 | NP_001107715.1 | 3 - 4 | 244 | 81 | |
XM_012956316.1 | XP_012811770.1 | 3 - 4 | 261 | 81 | ||
XM_012956317.2 | XP_012811771.1 | 3 - 4 | 299 | 81 | ||
GenBank mRNA | BC135301.1 | - | 2 - 3 | 229 | 81 | |
BC155970.1 | - | 3 - 4 | 244 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |