Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114261.2 | NP_001107733.2 | 1 - 2 | 178 | 80 | |
GenBank mRNA | CR443913.1 | - | 1 - 2 | 176 | 80 | |
CR443914.1 | - | 2 - 3 | 490 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |