Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114265.1 | NP_001107737.1 | 3 - 4 | 644 | 96 | |
XM_012954575.2 | XP_012810029.1 | 3 - 4 | 1079 | 96 | ||
XM_012954610.2 | XP_012810064.1 | 4 - 5 | 579 | 96 | ||
XM_012954644.1 | XP_012810098.1 | 4 - 5 | 451 | 96 | ||
XM_012954679.2 | XP_012810133.1 | 3 - 4 | 415 | 96 | ||
GenBank mRNA | BC158533.1 | - | 3 - 4 | 644 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |