Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114265.1 | NP_001107737.1 | 4 - 5 | 753 | 84 | |
XM_012954575.2 | XP_012810029.1 | 4 - 5 | 1188 | 84 | ||
XM_012954610.2 | XP_012810064.1 | 5 - 6 | 688 | 84 | ||
XM_012954644.1 | XP_012810098.1 | 5 - 6 | 560 | 84 | ||
XM_012954679.2 | XP_012810133.1 | 4 - 5 | 524 | 84 | ||
GenBank mRNA | BC158533.1 | - | 4 - 5 | 753 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |