Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114495.1 | NP_001107967.1 | 4 - 5 | 1370 | 67 | |
GenBank mRNA | BC159168.1 | - | 4 - 5 | 1370 | 67 | |
BC166202.1 | - | 4 - 5 | 2146 | 67 | ||
CT030340.1 | - | 4 - 5 | 2142 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |