Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122807.1 | NP_001116279.1 | 3 - 4 | 360 | 66 | |
GenBank mRNA | BC161222.1 | - | 3 - 4 | 360 | 66 | |
CU075789.1 | - | 3 - 4 | 353 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |