Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c2orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122809.1 | NP_001116281.1 | 2 - 3 | 964 | 81 | |
XM_012958482.2 | XP_012813936.1 | 3 - 4 | 1323 | 81 | ||
XM_012958483.2 | XP_012813937.1 | 4 - 5 | 1464 | 81 | ||
XM_012958484.2 | XP_012813938.1 | 3 - 4 | 1148 | 81 | ||
XM_012958485.2 | XP_012813939.1 | 3 - 4 | 605 | 81 | ||
XM_012958486.2 | XP_012813940.1 | 4 - 5 | 743 | 81 | ||
GenBank mRNA | BC152053.1 | - | 3 - 4 | 379 | 81 | |
BC161111.1 | - | 2 - 3 | 964 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |