Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c2orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122809.1 | NP_001116281.1 | 3 - 4 | 1058 | 67 | |
XM_012958482.2 | XP_012813936.1 | 4 - 5 | 67 | |||
XM_012958483.2 | XP_012813937.1 | 5 - 6 | 67 | |||
XM_012958484.2 | XP_012813938.1 | 4 - 5 | 67 | |||
XM_012958485.2 | XP_012813939.1 | 4 - 5 | 67 | |||
XM_012958486.2 | XP_012813940.1 | 5 - 6 | 67 | |||
GenBank mRNA | BC152053.1 | - | 4 - 5 | 67 | ||
BC161111.1 | - | 3 - 4 | 1058 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |