Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bves | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123416.1 | NP_001116888.1 | 8 - 9 | 1187 | 84 | |
GenBank mRNA | BC160594.1 | - | 8 - 9 | 1187 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |