Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123447.1 | NP_001116919.1 | 7 - 8 | 806 | 106 | |
XM_012960584.2 | XP_012816038.1 | 8 - 9 | 918 | 106 | ||
XM_012960585.2 | XP_012816039.1 | 8 - 9 | 1002 | 106 | ||
XM_018092916.1 | XP_017948405.1 | 9 - 10 | 1025 | 106 | ||
GenBank mRNA | BC161493.1 | - | 7 - 8 | 806 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |