Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il5ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123462.1 | NP_001116934.1 | 9 - 10 | 1227 | 101 | |
XM_012960586.2 | XP_012816040.1 | 9 - 10 | 1227 | 101 | ||
XM_012960587.2 | XP_012816041.1 | 10 - 11 | 1349 | 101 | ||
XM_012960588.1 | XP_012816042.1 | 9 - 10 | 1197 | 101 | ||
XM_012960589.2 | XP_012816043.1 | 9 - 10 | 1196 | 101 | ||
GenBank mRNA | BC161054.1 | - | 9 - 10 | 1227 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |