Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: march5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123478.1 | NP_001116950.1 | 2 - 3 | 411 | 74 | |
GenBank mRNA | BC160533.1 | - | 2 - 3 | 411 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |