Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126119.1 | NP_001119591.1 | 4 - 5 | 294 | 104 | |
GenBank mRNA | BC077708.1 | - | 4 - 5 | 257 | 104 | |
BC118887.1 | - | 3 - 4 | 281 | 104 | ||
BC135839.1 | - | 4 - 5 | 294 | 104 | ||
BC161316.1 | - | 3 - 4 | 268 | 104 | ||
CT025311.2 | - | 3 - 4 | 227 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |