Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126501.1 | NP_001119973.1 | 4 - 5 | 443 | 74 | |
GenBank mRNA | BC158190.1 | - | 4 - 5 | 443 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |