Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126556.1 | NP_001120028.1 | 6 - 7 | 733 | 80 | |
XM_018097039.1 | XP_017952528.1 | 6 - 7 | 767 | 80 | ||
GenBank mRNA | BC158333.1 | - | 6 - 7 | 733 | 80 | |
BC161197.1 | - | 6 - 7 | 704 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |