Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126738.1 | NP_001120210.1 | 5 - 6 | 555 | 82 | |
GenBank mRNA | BC159357.1 | - | 5 - 6 | 555 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |