Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankdd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127008.1 | NP_001120480.1 | 11 - 12 | 1296 | 75 | |
XM_012956326.2 | XP_012811780.1 | 11 - 12 | 1144 | 75 | ||
XM_018091063.1 | XP_017946552.1 | 11 - 12 | 1168 | 75 | ||
GenBank mRNA | BC161348.1 | - | 11 - 12 | 1296 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |