Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127012.1 | NP_001120484.1 | 1 - 2 | 831 | 65 | |
XM_012968029.2 | XP_012823483.1 | 2 - 3 | 530 | 65 | ||
XM_012968031.2 | XP_012823485.1 | 2 - 3 | 542 | 65 | ||
GenBank mRNA | BC161354.1 | - | 1 - 2 | 831 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |