Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127022.1 | NP_001120494.1 | 1 - 2 | 48 | 65 | |
GenBank mRNA | BC161373.1 | - | 1 - 2 | 48 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |