Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kif19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012959642.2 | XP_012815096.1 | 6 - 7 | 1092 | 83 | |
GenBank mRNA | BC161478.1 | - | 781 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |