Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127127.1 | NP_001120599.1 | 3 - 4 | 356 | 88 | |
GenBank mRNA | AB474592.1 | - | 2 - 3 | 262 | 88 | |
BC161634.1 | - | 3 - 4 | 356 | 88 | ||
CT030302.1 | - | 3 - 4 | 309 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |