Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pigb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127136.1 | NP_001120608.1 | 9 - 10 | 1207 | 65 | |
XM_018091940.1 | XP_017947429.1 | 9 - 10 | 1105 | 65 | ||
GenBank mRNA | BC161737.1 | - | 9 - 10 | 1207 | 65 | |
CT025216.2 | - | 9 - 10 | 1383 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |