Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abhd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127147.1 | NP_001120619.1 | 6 - 7 | 828 | 94 | |
GenBank mRNA | BC161759.1 | - | 6 - 7 | 828 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |