Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: capza1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017324.3 | NP_001017324.1 | 9 - 10 | 798 | 103 | |
GenBank mRNA | BC067322.1 | - | 727 | 103 | ||
BC158140.1 | - | 9 - 10 | 798 | 103 | ||
CR848613.2 | - | 9 - 10 | 746 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |