Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: carnmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126522.1 | NP_001119994.1 | 6 - 7 | 1314 | 137 | |
GenBank mRNA | BC158255.1 | - | 6 - 7 | 1314 | 137 | |
CU024918.1 | - | 6 - 7 | 939 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |