Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126533.1 | NP_001120005.1 | 2 - 3 | 239 | 84 | |
GenBank mRNA | BC158290.1 | - | 2 - 3 | 239 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |