Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126533.1 | NP_001120005.1 | 9 - 10 | 1090 | 96 | |
GenBank mRNA | BC158290.1 | - | 9 - 10 | 1090 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |