Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b4galt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126545.1 | NP_001120017.1 | 5 - 6 | 1378 | 67 | |
XM_012958539.2 | XP_012813993.1 | 5 - 6 | 854 | 67 | ||
XM_012958540.1 | XP_012813994.1 | 5 - 6 | 814 | 67 | ||
GenBank mRNA | BC158315.1 | - | 5 - 6 | 1378 | 67 | |
CT025270.2 | - | 5 - 6 | 862 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |