Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gid4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126573.1 | NP_001120045.1 | 3 - 4 | 505 | 64 | |
GenBank mRNA | BC158383.1 | - | 3 - 4 | 505 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |