Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: utp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126577.1 | NP_001120049.1 | 15 - 16 | 1325 | 93 | |
XM_012971526.1 | XP_012826980.1 | 15 - 16 | 1426 | 93 | ||
GenBank mRNA | BC158387.1 | - | 15 - 16 | 1325 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |