Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: utp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126577.1 | NP_001120049.1 | 6 - 7 | 402 | 123 | |
XM_012971526.1 | XP_012826980.1 | 6 - 7 | 503 | 123 | ||
GenBank mRNA | BC158387.1 | - | 6 - 7 | 402 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |