Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126759.1 | NP_001120231.1 | 8 - 9 | 661 | 69 | |
GenBank mRNA | BC160408.1 | - | 8 - 9 | 661 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |