Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126769.1 | NP_001120241.1 | 2 - 3 | 396 | 97 | |
GenBank mRNA | BC160433.1 | - | 2 - 3 | 396 | 97 | |
BC161596.1 | - | 2 - 3 | 346 | 97 | ||
CT025214.2 | - | 2 - 3 | 279 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |