Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126779.1 | NP_001120251.1 | 2 - 3 | 339 | 83 | |
GenBank mRNA | BC160467.1 | - | 2 - 3 | 339 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |