Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plppr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126788.1 | NP_001120260.1 | 2 - 3 | 157 | 65 | |
GenBank mRNA | BC160501.1 | - | 2 - 3 | 157 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |