Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126809.1 | NP_001120281.1 | 4 - 5 | 857 | 97 | |
GenBank mRNA | BC160546.1 | - | 4 - 5 | 857 | 97 | |
CU025145.1 | - | 5 - 6 | 908 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |