Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126809.1 | NP_001120281.1 | 6 - 7 | 1295 | 90 | |
GenBank mRNA | BC160546.1 | - | 6 - 7 | 1295 | 90 | |
CU025145.1 | - | 7 - 8 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |