Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnnm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126912.2 | NP_001120384.2 | 2 - 3 | 1708 | 78 | |
XM_012966082.2 | XP_012821536.1 | 2 - 3 | 2340 | 78 | ||
XM_012966083.2 | XP_012821537.1 | 2 - 3 | 2341 | 78 | ||
XM_012966084.2 | XP_012821538.1 | 2 - 3 | 2341 | 78 | ||
GenBank mRNA | BC161088.1 | - | 2 - 3 | 1708 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |