Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126926.1 | NP_001120398.1 | 3 - 4 | 822 | 100 | |
XM_018092927.1 | XP_017948416.1 | 3 - 4 | 816 | 100 | ||
XM_018092928.1 | XP_017948417.1 | 3 - 4 | 931 | 100 | ||
GenBank mRNA | BC161108.1 | - | 3 - 4 | 822 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |