Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126935.1 | NP_001120407.1 | 4 - 5 | 593 | 100 | |
XM_012962732.2 | XP_012818186.1 | 4 - 5 | 673 | 100 | ||
XM_012962733.2 | XP_012818187.1 | 4 - 5 | 739 | 100 | ||
XM_012962734.2 | XP_012818188.1 | 4 - 5 | 637 | 100 | ||
XM_012962735.2 | XP_012818189.1 | 5 - 6 | 1144 | 100 | ||
XM_012962736.2 | XP_012818190.1 | 5 - 6 | 1141 | 100 | ||
XM_012962737.1 | XP_012818191.1 | 4 - 5 | 630 | 100 | ||
XM_012962738.2 | XP_012818192.2 | 2 - 3 | 206 | 100 | ||
GenBank mRNA | BC161123.1 | - | 4 - 5 | 593 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |