Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rimkla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126982.1 | NP_001120454.1 | 577 | 69 | ||
GenBank mRNA | BC161236.1 | - | 577 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |