Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hypk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127003.1 | NP_001120475.1 | 3 - 4 | 271 | 73 | |
GenBank mRNA | BC161322.1 | - | 3 - 4 | 271 | 73 | |
CR942732.2 | - | 3 - 4 | 268 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |