Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127052.1 | NP_001120524.1 | 1 - 2 | 377 | 87 | |
XM_018092936.1 | XP_017948425.1 | 1 - 2 | 538 | 87 | ||
GenBank mRNA | BC124565.1 | - | 1 - 2 | 377 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |