Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pyroxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127086.1 | NP_001120558.1 | 1962 | 83 | ||
GenBank mRNA | BC161508.1 | - | 1962 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis |