Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127090.1 | NP_001120562.1 | 201 | 69 | ||
GenBank mRNA | BC161518.1 | - | 201 | 69 | ||
CT025325.2 | - | 2 - 3 | 177 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |