Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127168.1 | NP_001120640.1 | 9 - 10 | 1871 | 79 | |
XM_012958613.2 | XP_012814067.1 | 10 - 11 | 2036 | 79 | ||
XM_012958614.2 | XP_012814068.1 | 10 - 11 | 1962 | 79 | ||
GenBank mRNA | BC166189.1 | - | 9 - 10 | 1871 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |