Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: maats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127421.1 | NP_001120893.1 | 3 - 4 | 382 | 82 | |
GenBank mRNA | BC166178.1 | - | 3 - 4 | 382 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |