Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retsat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127940.1 | NP_001121412.1 | 1 - 2 | 184 | 64 | |
XM_012958621.2 | XP_012814075.1 | 2 - 3 | 389 | 64 | ||
XM_012958622.1 | XP_012814076.1 | 2 - 3 | 388 | 64 | ||
GenBank mRNA | BC166112.1 | - | 1 - 2 | 184 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |