Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retsat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127940.1 | NP_001121412.1 | 2 - 3 | 361 | 83 | |
XM_012958621.2 | XP_012814075.1 | 3 - 4 | 566 | 83 | ||
XM_012958622.1 | XP_012814076.1 | 3 - 4 | 565 | 83 | ||
GenBank mRNA | BC166112.1 | - | 2 - 3 | 361 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |