Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: siglec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127949.1 | NP_001121421.1 | 2504 | 104 | ||
GenBank mRNA | BC166151.1 | - | 2504 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |